ENA065085580 Enamine

methyl 4-(1,1'-biphenyl-4-yl)-2,4-dioxobutanoate

  • MDL number MFCD07346275
  • Cas Number : 63656-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14629

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14629
Availability details for
Quantity
  • Availability
  • Add to Cart