ENA065085581 Enamine

2-[(propylsulfonyl)amino]propanoic acid

  • MDL number MFCD07345610
  • Cas Number : 1007984-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14630

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14630
Availability details for
Quantity
  • Availability
  • Add to Cart