ENA065085587 Enamine

(5-thien-3-yl-2H-tetrazol-2-yl)acetic acid

  • MDL number MFCD07345668
  • Cas Number : 869472-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14641

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14641
Availability details for
Quantity
  • Availability
  • Add to Cart