ENA065085595 Enamine

4-(diethylsulfamoyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD07345670
  • Cas Number : 869472-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-251
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14661
Availability details for
Quantity
  • Availability
  • Add to Cart