ENA065085598 Enamine

4-(morpholine-4-sulfonyl)-1H-pyrrole-2-carbohydrazide

  • MDL number MFCD07346305
  • Cas Number : 870692-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14668
Availability details for
Quantity
  • Availability
  • Add to Cart