ENA065085607 Enamine

(2E)-3-(2-ethyl-1-benzofuran-3-yl)acrylic acid

  • MDL number MFCD07345517
  • Cas Number : 91902-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14688

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14688
Availability details for
Quantity
  • Availability
  • Add to Cart