ENA065085615 Enamine

2-methoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]aniline

  • MDL number MFCD07346330
  • Cas Number : 870692-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14699
Availability details for
Quantity
  • Availability
  • Add to Cart