ENA065085626 Enamine

3-(2-chloroacetyl)-1-(2,2,2-trifluoroethyl)urea

  • MDL number MFCD07346326
  • Cas Number : 870692-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14712
Availability details for
Quantity
  • Availability
  • Add to Cart