ENA065085629 Enamine

2-hydroxy-3-[4-(propan-2-yl)phenyl]-3,4-dihydroquinazolin-4-one

  • MDL number MFCD07346331
  • Molecular Weight : 280.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14718
Availability details for
Quantity
  • Availability
  • Add to Cart