ENA065085631 Enamine

4-fluoro-7-hydroxy-3-methyl-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD07346352
  • Cas Number : 870693-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14721
Availability details for
Quantity
  • Availability
  • Add to Cart