ENA065085634 Enamine

(1-cyclopropylethyl)[(4-methoxyphenyl)methyl]amine

  • MDL number MFCD07348573
  • Cas Number : 871217-49-1
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14732
Availability details for
Quantity
  • Availability
  • Add to Cart