ENA065085638 Enamine

1-(chloroacetyl)-4-(2,3-dihydro-1H-inden-5-ylsulfonyl)piperazine

  • MDL number MFCD07348596
  • Cas Number : 871217-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120...122
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14741

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14741
Availability details for
Quantity
  • Availability
  • Add to Cart