ENA065085640 Enamine

2-(chloroacetyl)decahydroisoquinoline

  • MDL number MFCD07348574
  • Cas Number : 871217-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217...219
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14743

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14743
Availability details for
Quantity
  • Availability
  • Add to Cart