ENA065085654 Enamine

1-(2-cyanobenzenesulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD07348753
  • Cas Number : 871478-83-0
  • Molecular Weight : 294.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14765
Availability details for
Quantity
  • Availability
  • Add to Cart