ENA065085660 Enamine

methyl({[2-(pyrrolidin-1-yl)phenyl]methyl})amine

  • MDL number MFCD07348555
  • Cas Number : 871217-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14773
Availability details for
Quantity
  • Availability
  • Add to Cart