ENA065085664 Enamine

5-[(dimethylamino)methyl]-4-(4-methylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD07346344
  • Cas Number : 870693-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-164
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14783
Availability details for
Quantity
  • Availability
  • Add to Cart