ENA065085670 Enamine

6-amino-1-ethyl-5-(2-methyl-1,3-thiazol-4-yl)pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD07348562
  • Cas Number : 871217-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250...252
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14798

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14798
Availability details for
Quantity
  • Availability
  • Add to Cart