ENA065085673 Enamine

4-[(5,6,7,8-tetrahydronaphthalen-2-ylsulfonyl)amino]butanoic acid

  • MDL number MFCD07346353
  • Cas Number : 870693-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14802

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14802
Availability details for
Quantity
  • Availability
  • Add to Cart