ENA065085677 Enamine

2-methyl-4-phenyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD07348569
  • Cas Number : 32002-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14811

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14811
Availability details for
Quantity
  • Availability
  • Add to Cart