ENA065085678 Enamine

2-[(2,4-dimethyl-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazin-6-yl)thio]acetohydrazide

  • MDL number MFCD07346356
  • Cas Number : 870693-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14812

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14812
Availability details for
Quantity
  • Availability
  • Add to Cart