ENA065085683 Enamine

6-(2-mercapto-4-oxoquinazolin-3(4H)-yl)hexanamide

  • MDL number MFCD07366409
  • Cas Number : 871478-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 261...263
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14824

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14824
Availability details for
Quantity
  • Availability
  • Add to Cart