ENA065085690 Enamine

2-sulfanyl-3-(4H-1,2,4-triazol-3-yl)-3,4-dihydroquinazolin-4-one

  • MDL number MFCD07366415
  • Cas Number : 131842-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234-235
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14833
Availability details for
Quantity
  • Availability
  • Add to Cart