ENA065085692 Enamine

4-(1H-1,2,3-benzotriazol-1-ylmethyl)benzoic acid

  • MDL number MFCD07348585
  • Cas Number : 502649-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 264...266
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14835

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14835
Availability details for
Quantity
  • Availability
  • Add to Cart