ENA065085695 Enamine

2-(4-oxo-1,4-dihydrocinnolin-1-yl)acetic acid

  • MDL number MFCD07348742
  • Cas Number : 203252-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14839
Availability details for
Quantity
  • Availability
  • Add to Cart