ENA065085703 Enamine

6-methyl-2-oxo-4-thioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD07348579
  • Cas Number : 2228-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14852

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14852
Availability details for
Quantity
  • Availability
  • Add to Cart