ENA065085706 Enamine

5-acetyl-6-methyl-4-sulfanyl-1,2-dihydropyrimidin-2-one

  • MDL number MFCD26143644
  • Cas Number : 90151-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14862
Availability details for
Quantity
  • Availability
  • Add to Cart