ENA065085708 Enamine

2-[3,5-dimethyl-1-(2-methylpropyl)-1H-pyrazol-4-yl]acetic acid

  • MDL number MFCD07348595
  • Cas Number : 957023-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14864
Availability details for
Quantity
  • Availability
  • Add to Cart