ENA065085717 Enamine

2-[5-(phenoxymethyl)-2H-1,2,3,4-tetrazol-2-yl]acetic acid

  • MDL number MFCD07352283
  • Cas Number : 871497-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-112
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14877
Availability details for
Quantity
  • Availability
  • Add to Cart