ENA065085724 Enamine

6-chloro-3,4-dihydro-2H-1-benzopyran-3-carboxylic acid

  • MDL number MFCD04114612
  • Cas Number : 164265-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-145
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14886
Availability details for
Quantity
  • Availability
  • Add to Cart