ENA065085729 Enamine

1-(4-ethylbenzyl)-6-methyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD07352289
  • Cas Number : 871497-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14893
Availability details for
Quantity
  • Availability
  • Add to Cart