ENA065085731 Enamine

2-chloro-N-(1-cyanocyclohexyl)-N-methylpropanamide

  • MDL number MFCD07352294
  • Cas Number : 871497-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14896

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14896
Availability details for
Quantity
  • Availability
  • Add to Cart