ENA065085732 Enamine

4-(aminosulfonyl)benzenesulfonyl chloride

  • MDL number MFCD07352256
  • Cas Number : 46249-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14899

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-14899
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart