ENA065085740 Enamine

2-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)-2-oxoacetic acid

  • MDL number MFCD07352302
  • Cas Number : 956438-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14911
Availability details for
Quantity
  • Availability
  • Add to Cart