ENA065085756 Enamine

6-methyl-1-[(4-methylphenyl)methyl]-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD07352244
  • Cas Number : 871548-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-109
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14951
Availability details for
Quantity
  • Availability
  • Add to Cart