ENA065085777 Enamine

3-(2-chloroacetyl)-1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)urea

  • MDL number MFCD07339184
  • Cas Number : 863669-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14283
Availability details for
Quantity
  • Availability
  • Add to Cart