ENA065085797 Enamine

4-(morpholine-4-sulfonyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD07345594
  • Cas Number : 869464-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-241
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14621
Availability details for
Quantity
  • Availability
  • Add to Cart