ENA065085820 Enamine

2-[(3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazin-6-yl)sulfanyl]acetohydrazide

  • MDL number MFCD00690973
  • Cas Number : 329709-86-6
  • Molecular Weight : 217.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-226
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14644
Availability details for
Quantity
  • Availability
  • Add to Cart