ENA065085827 Enamine

3-[2-(4-fluorophenyl)-1H-indol-3-yl]propanoic acid

  • MDL number MFCD06660846
  • Cas Number : 869472-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-121
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14651
Availability details for
Quantity
  • Availability
  • Add to Cart