ENA065085828 Enamine

3-amino-N-benzyl-3-thioxopropanamide

  • MDL number MFCD04606717
  • Cas Number : 102817-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14652

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14652
Availability details for
Quantity
  • Availability
  • Add to Cart