ENA065085846 Enamine

methyl (6-oxo-2-thioxo-1,2,3,6-tetrahydropyrimidin-4-yl)acetate

  • MDL number MFCD07343862
  • Cas Number : 868238-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185...187
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14423

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14423
Availability details for
Quantity
  • Availability
  • Add to Cart