ENA065085856 Enamine

3-(6-chloro-2-oxo-2,3-dihydro-1,3-benzoxazol-3-yl)propanoic acid

  • MDL number MFCD07339245
  • Cas Number : 17124-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14302
Availability details for
Quantity
  • Availability
  • Add to Cart