ENA065085861 Enamine

(4-oxo-3,4-dihydroquinazolin-2-yl)acetonitrile

  • MDL number MFCD00481615
  • Cas Number : 30750-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244...246
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14238

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14238
Availability details for
Quantity
  • Availability
  • Add to Cart