ENA065085862 Enamine

5-amino-3-methyl-1,2-thiazole-4-carboxylic acid

  • MDL number MFCD06376749
  • Cas Number : 22131-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14305
Availability details for
Quantity
  • Availability
  • Add to Cart