ENA065085865 Enamine

2-anilino-4-oxo-4,5-dihydrofuran-3-carboxylic acid

  • MDL number MFCD00877764
  • Cas Number : 58337-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14432

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14432
Availability details for
Quantity
  • Availability
  • Add to Cart