ENA065085878 Enamine

4-[(ethoxycarbonyl)amino]benzoic acid

  • MDL number MFCD00610504
  • Cas Number : 5180-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14441

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14441
Availability details for
Quantity
  • Availability
  • Add to Cart