ENA065085890 Enamine

5-(4-tert-butylphenyl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD05182332
  • Cas Number : 20586-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213...215
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14714

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14714
Availability details for
Quantity
  • Availability
  • Add to Cart