ENA065085898 Enamine

2-(chloromethyl)-5-(2-chlorophenyl)-1,3,4-oxadiazole

  • MDL number MFCD07366400
  • Cas Number : 24023-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91...93
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14723

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14723
Availability details for
Quantity
  • Availability
  • Add to Cart