ENA065085903 Enamine

2-chloro-N-[2-(1H-indol-3-yl)ethyl]acetamide

  • MDL number MFCD00120131
  • Cas Number : 52191-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14456

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14456
Availability details for
Quantity
  • Availability
  • Add to Cart