ENA065085913 Enamine

2-(chloroacetyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD00264807
  • Cas Number : 111631-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231...233
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14739

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14739
Availability details for
Quantity
  • Availability
  • Add to Cart