ENA065085931 Enamine

2-(thiophen-3-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD07343850
  • Cas Number : 868238-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14475
Availability details for
Quantity
  • Availability
  • Add to Cart